Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cul5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_143408.3 | NP_651665.2 | 2 - 3 | 1702 | 91 | |
GenBank mRNA | BT133427.1 | - | 2 - 3 | 1886 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |