Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTA1-like | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275364.2 | NP_001262293.1 | 2 - 3 | 410 | 57 | |
NM_169086.3 | NP_730976.1 | 2 - 3 | 410 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |