Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170148.2 | NP_001163619.1 | 2 - 3 | 206 | 79 | |
NM_001260190.1 | NP_001247119.1 | 2 - 3 | 224 | 79 | ||
NM_001275662.1 | NP_001262591.1 | 2 - 3 | 206 | 79 | ||
NM_001382175.1 | NP_001369014.1 | 2 - 3 | 493 | 79 | ||
NM_169633.5 | NP_732001.3 | 2 - 3 | 456 | 79 | ||
NM_169634.3 | NP_732002.1 | 2 - 3 | 206 | 79 | ||
NM_169635.3 | NP_732003.1 | 2 - 3 | 639 | 79 | ||
NM_169636.3 | NP_732004.1 | 2 - 3 | 201 | 79 | ||
NM_169637.4 | NP_732005.2 | 2 - 3 | 224 | 79 | ||
NM_169638.4 | NP_732006.2 | 2 - 3 | 206 | 79 | ||
NM_206494.3 | NP_996216.1 | 2 - 3 | 206 | 79 | ||
NM_206495.2 | NP_996217.1 | 2 - 3 | 224 | 79 | ||
GenBank mRNA | BT001534.1 | - | 2 - 3 | 204 | 79 | |
BT011469.1 | - | 2 - 3 | 204 | 79 | ||
BT133132.1 | - | 2 - 3 | 176 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster |