Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mical | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260085.2 | NP_001247014.1 | 15 - 16 | 9382 | 72 | |
NM_001260086.3 | NP_001247015.1 | 15 - 16 | 9430 | 72 | ||
NM_001260087.3 | NP_001247016.2 | 15 - 16 | 9670 | 72 | ||
NM_176445.2 | NP_788622.1 | 12 - 13 | 9275 | 72 | ||
NM_176446.2 | NP_788623.1 | 11 - 12 | 9231 | 72 | ||
NM_176447.2 | NP_788624.1 | 15 - 16 | 9397 | 72 | ||
NM_176448.2 | NP_788625.1 | 12 - 13 | 9171 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |