Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mgat2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014684.2 | NP_001014684.1 | 4 - 5 | 638 | 79 | |
NM_001276167.1 | NP_001263096.1 | 4 - 5 | 937 | 79 | ||
NM_143506.4 | NP_651763.4 | 4 - 5 | 937 | 79 | ||
GenBank mRNA | AB176553.1 | - | 3 - 4 | 292 | 79 | |
AY047566.1 | - | 4 - 5 | 903 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |