Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CrebB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103546.3 | NP_001097016.2 | 5 - 6 | 1115 | 90 | |
NM_001103548.4 | NP_001097018.1 | 4 - 5 | 1357 | 90 | ||
NM_001272749.2 | NP_001259678.1 | 5 - 6 | 1073 | 90 | ||
NM_206781.4 | NP_996504.1 | 4 - 5 | 1096 | 90 | ||
GenBank mRNA | BT028813.2 | - | 3 - 4 | 664 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Drosophila melanogaster |