Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ApepP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259124.2 | NP_001246053.1 | 1 | 2068 | 85 | |
NM_058061.5 | NP_477409.1 | 2 | 1944 | 85 | ||
GenBank mRNA | AY061297.1 | - | 2 | 1957 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster |