Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kdm5bb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002166.2 | NP_001002166.2 | 1 - 2 | 324 | 59 | |
XM_005167024.4 | XP_005167081.1 | 1 - 2 | 945 | 59 | ||
XM_009303989.3 | XP_009302264.1 | 1 - 2 | 945 | 59 | ||
GenBank mRNA | BC071280.1 | - | 1 - 2 | 257 | 59 | |
EH449360.1 | - | 1 - 2 | 334 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |