Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ptn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003981.2 | NP_001003981.2 | 3 - 4 | 427 | 65 | |
GenBank mRNA | AY572239.1 | - | 3 - 4 | 358 | 65 | |
BC095549.1 | - | 3 - 4 | 427 | 65 | ||
BC165895.1 | - | 2 - 3 | 294 | 65 | ||
KM655586.1 | - | 2 - 3 | 260 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |