Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mapk6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045552.1 | NP_001039017.1 | 4 - 5 | 1066 | 123 | |
XM_017351949.2 | XP_017207438.1 | 5 - 6 | 1842 | 123 | ||
XM_017351950.2 | XP_017207439.1 | 5 - 6 | 2018 | 123 | ||
XM_017351951.2 | XP_017207440.1 | 5 - 6 | 1860 | 123 | ||
GenBank mRNA | BC162744.1 | - | 4 - 5 | 1153 | 123 | |
DQ360074.1 | - | 4 - 5 | 1066 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |