Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: baiap2l1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198922.1 | NP_944604.1 | 6 - 7 | 640 | 72 | |
XM_009306593.3 | XP_009304868.1 | 6 - 7 | 679 | 72 | ||
GenBank mRNA | AY398378.1 | - | 6 - 7 | 641 | 72 | |
BC050238.1 | - | 6 - 7 | 640 | 72 | ||
BC068330.1 | - | 6 - 7 | 624 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |