Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rbm42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003850.1 | NP_001003850.1 | 9 - 10 | 1072 | 67 | |
GenBank mRNA | AY648798.1 | - | 9 - 10 | 1072 | 67 | |
BC095046.1 | - | 9 - 10 | 1040 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |