Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fance | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040634.1 | NP_001035724.1 | 3 - 4 | 812 | 73 | |
GenBank mRNA | AY968595.1 | - | 3 - 4 | 812 | 73 | |
BC162341.1 | - | 3 - 4 | 835 | 73 | ||
BC162347.1 | - | 3 - 4 | 835 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |