Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nfyba | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017565.1 | NP_001017565.1 | 4 - 5 | 449 | 68 | |
XM_005168523.4 | XP_005168580.1 | 4 - 5 | 445 | 68 | ||
GenBank mRNA | BC092926.1 | - | 4 - 5 | 449 | 68 | |
BC164291.1 | - | 3 - 4 | 454 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |