Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cpsf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131009.1 | NP_571084.1 | 5 - 6 | 794 | 57 | |
XM_021469299.1 | XP_021324974.1 | 5 - 6 | 829 | 57 | ||
GenBank mRNA | BC045289.1 | - | 5 - 6 | 797 | 57 | |
BC164456.1 | - | 5 - 6 | 535 | 57 | ||
U70479.1 | - | 5 - 6 | 794 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |