Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fhl2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006028.2 | NP_001006028.2 | 5 - 6 | 771 | 95 | |
GenBank mRNA | BC083393.1 | - | 5 - 6 | 776 | 95 | |
DQ118097.1 | - | 4 - 5 | 700 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |