Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bhlhe41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039107.1 | NP_001034196.1 | 3 - 4 | 255 | 81 | |
GenBank mRNA | AB212760.1 | - | 3 - 4 | 255 | 81 | |
BC163162.1 | - | 3 - 4 | 255 | 81 | ||
BC163191.1 | - | 3 - 4 | 255 | 81 | ||
DQ779789.1 | - | 3 - 4 | 315 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |