Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pdpk1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077344.1 | NP_001070812.1 | 10 - 11 | 1373 | 118 | |
GenBank mRNA | BC124736.1 | - | 10 - 11 | 1373 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |