Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mdh1aa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001309523.2 | NP_001296452.1 | 4 - 5 | 775 | 77 | |
NM_001316925.1 | NP_001303854.1 | 4 - 5 | 488 | 77 | ||
NM_199947.3 | NP_956241.2 | 4 - 5 | 488 | 77 | ||
GenBank mRNA | AY161294.1 | - | 4 - 5 | 424 | 77 | |
AY391437.1 | - | 4 - 5 | 414 | 77 | ||
BC053158.1 | - | 4 - 5 | 411 | 77 | ||
EH453718.1 | - | 4 - 5 | 465 | 77 | ||
EH579651.1 | - | 4 - 5 | 596 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |