Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ppdpfa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200079.1 | NP_956373.1 | 3 - 4 | 325 | 83 | |
GenBank mRNA | BC057474.1 | - | 3 - 4 | 325 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |