Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gar1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200975.2 | NP_957269.2 | 4 - 5 | 513 | 157 | |
GenBank mRNA | BC045900.1 | - | 4 - 5 | 486 | 157 | |
EH495404.1 | - | 4 - 5 | 542 | 157 | ||
EH520414.1 | - | 3 - 4 | 767 | 156 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |