Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fbp1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213132.1 | NP_998297.1 | 5 - 6 | 763 | 87 | |
GenBank mRNA | BC053267.1 | - | 5 - 6 | 763 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Probe spans exons |