Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ctdspla | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045447.2 | NP_001038912.2 | 6 - 7 | 1017 | 67 | |
XM_005162710.4 | XP_005162767.1 | 7 - 8 | 1056 | 67 | ||
GenBank mRNA | BC122182.1 | - | 5 - 6 | 655 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |