Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pdgfab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076757.1 | NP_001070225.1 | 6 - 7 | 1231 | 62 | |
GenBank mRNA | BC124288.1 | - | 6 - 7 | 1231 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |