Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: es1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131039.1 | NP_571114.1 | 2 - 3 | 228 | 59 | |
GenBank mRNA | BC122433.1 | - | 2 - 3 | 209 | 59 | |
U10403.1 | - | 2 - 3 | 228 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |