Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dachb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_152956.2 | NP_694488.2 | 7 - 8 | 1318 | 99 | |
XM_009305776.3 | XP_009304051.1 | 7 - 8 | 1346 | 99 | ||
XM_021477542.1 | XP_021333217.1 | 6 - 7 | 731 | 99 | ||
GenBank mRNA | AF427109.1 | - | 7 - 8 | 1292 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Danio rerio Probe spans exons |