Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hm13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212572.2 | NP_997737.2 | 10 - 11 | 1090 | 80 | |
XM_005161890.4 | XP_005161947.1 | 10 - 11 | 1127 | 80 | ||
GenBank mRNA | AY926458.1 | - | 10 - 11 | 1056 | 80 | |
BC053414.1 | - | 10 - 11 | 1027 | 80 | ||
BC071503.1 | - | 10 - 11 | 1090 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |