Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: calhm2.1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003458.1 | NP_001003458.1 | 1 - 2 | 750 | 114 | |
XM_005156784.4 | XP_005156841.1 | 2 - 3 | 832 | 114 | ||
GenBank mRNA | BC078392.1 | - | 1 - 2 | 750 | 114 | |
BC164090.1 | - | 1 - 2 | 581 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Probe spans exons |