Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mtdha | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007135.2 | NP_001007136.2 | 8 - 9 | 1287 | 102 | |
XM_005159603.3 | XP_005159660.1 | 8 - 9 | 1363 | 102 | ||
XM_005159604.3 | XP_005159661.1 | 8 - 9 | 1364 | 102 | ||
XM_017352195.2 | XP_017207684.1 | 8 - 9 | 1367 | 102 | ||
GenBank mRNA | BC068430.1 | - | 8 - 9 | 1287 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |