Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tspan15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017773.1 | NP_001017773.1 | 5 - 6 | 668 | 66 | |
GenBank mRNA | BC092907.1 | - | 5 - 6 | 668 | 66 | |
BC152126.1 | - | 5 - 6 | 577 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |