Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: acsf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045419.2 | NP_001038884.2 | 16 - 17 | 1973 | 107 | |
NM_001139438.1 | NP_001132910.1 | 15 - 16 | 2133 | 107 | ||
GenBank mRNA | BC122098.1 | - | 16 - 17 | 1968 | 107 | |
EH575812.1 | - | 1 - 2 | 654 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |