Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tefa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131400.1 | NP_571475.1 | 1 - 2 | 282 | 66 | |
XM_005156135.3 | XP_005156192.1 | 1 - 2 | 473 | 66 | ||
GenBank mRNA | BC078287.1 | - | 1 - 2 | 314 | 66 | |
U43671.1 | - | 1 - 2 | 282 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |