Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tfr1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009918.2 | NP_001009918.2 | 15 - 16 | 1847 | 96 | |
GenBank mRNA | AY649364.1 | - | 14 - 15 | 1687 | 96 | |
BC108025.1 | - | 15 - 16 | 1847 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |