Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gstp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001020513.1 | NP_001018349.1 | 4 - 5 | 288 | 68 | |
GenBank mRNA | AB194128.1 | - | 4 - 5 | 288 | 68 | |
BC163150.1 | - | 4 - 5 | 275 | 68 | ||
BC163154.1 | - | 4 - 5 | 275 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |