Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: jam3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082863.3 | NP_001076332.2 | 6 - 7 | 788 | 69 | |
GenBank mRNA | BC134233.1 | - | 6 - 7 | 788 | 69 | |
CU458901.1 | - | 6 - 7 | 695 | 69 | ||
CU458938.1 | - | 6 - 7 | 711 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |