Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vil1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200238.1 | NP_956532.1 | 17 - 18 | 2247 | 80 | |
XM_005167920.4 | XP_005167977.1 | 17 - 18 | 2325 | 80 | ||
GenBank mRNA | BC047186.1 | - | 17 - 18 | 2247 | 80 | |
BC165692.1 | - | 16 - 17 | 2200 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |