Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: m6pr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213205.1 | NP_998370.1 | 5 - 6 | 801 | 106 | |
GenBank mRNA | BC065858.1 | - | 5 - 6 | 801 | 106 | |
DQ089037.1 | - | 5 - 6 | 801 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |