Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pfkma | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004575.2 | NP_001004575.1 | 15 - 16 | 1587 | 102 | |
GenBank mRNA | BC081652.1 | - | 15 - 16 | 1563 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |