Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: il10rb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034185.3 | NP_001029357.3 | 2 - 3 | 220 | 78 | |
XM_005169113.3 | XP_005169170.1 | 3 - 4 | 296 | 78 | ||
XM_009304372.3 | XP_009302647.1 | 3 - 4 | 221 | 78 | ||
XM_009304374.3 | XP_009302649.1 | 2 - 3 | 240 | 78 | ||
XM_021478759.1 | XP_021334434.1 | 3 - 4 | 298 | 78 | ||
XM_021478760.1 | XP_021334435.1 | 2 - 3 | 144 | 78 | ||
GenBank mRNA | AM039829.1 | - | 2 - 3 | 181 | 78 | |
BC124745.1 | - | 2 - 3 | 196 | 78 | ||
BC164411.1 | - | 2 - 3 | 204 | 78 | ||
DQ198217.1 | - | 2 - 3 | 230 | 78 | ||
DT864428.1 | - | 2 - 3 | 220 | 78 | ||
EF014955.1 | - | 2 - 3 | 236 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Danio rerio Inventoried Probe spans exons |