Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mylk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110520.2 | NP_001103990.1 | 3 - 4 | 954 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Danio rerio Probe spans exons |