Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: casp3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131877.3 | NP_571952.1 | 5 - 6 | 772 | 66 | |
GenBank mRNA | AB047003.1 | - | 4 - 5 | 491 | 66 | |
BC078310.1 | - | 5 - 6 | 776 | 66 | ||
KX084794.1 | - | 4 - 5 | 491 | 66 | ||
MG957994.1 | - | 4 - 5 | 491 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |