Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fgfr1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001309399.1 | NP_001296328.1 | 8 - 9 | 1697 | 67 | |
NM_152962.3 | NP_694494.2 | 8 - 9 | 1697 | 67 | ||
GenBank mRNA | AF389400.2 | - | 7 - 8 | 1079 | 67 | |
AY197498.1 | - | 7 - 8 | 1079 | 67 | ||
BC162342.1 | - | 7 - 8 | 1079 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |