Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hes6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_194400.2 | NP_919381.2 | 2 - 3 | 227 | 61 | |
GenBank mRNA | AB158461.1 | - | 2 - 3 | 210 | 61 | |
BC062387.1 | - | 2 - 3 | 214 | 61 | ||
BC065599.1 | - | 2 - 3 | 227 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |