Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rps6kal | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200073.1 | NP_956367.1 | 18 - 19 | 1870 | 65 | |
XM_009307718.2 | XP_009305993.1 | 13 - 14 | 1367 | 65 | ||
XM_021480984.1 | XP_021336659.1 | 11 - 12 | 1175 | 65 | ||
GenBank mRNA | BC045478.1 | - | 18 - 19 | 1975 | 65 | |
BC057467.1 | - | 18 - 19 | 1870 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |