Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fancb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040636.1 | NP_001035726.1 | 1 - 2 | 919 | 67 | |
GenBank mRNA | AY968593.1 | - | 1 - 2 | 919 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Danio rerio Probe spans exons |