Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
TaqMan
®
Gene Expression Assays
›
Search Tool
›
Search Results
› Dr03136902_m1
See other NRXN1A GE Assays ›
Gene Symbol:
nrxn1a
Gene Name:
neurexin 1a
Gene Aliases:
-
Chromosome Location:
Chr.12: 114926 - 403053 on Build GRCz11
Species:
Zebrafish
Species Specific ID (Flybase ID):
-
Assay ID
Dr03136902_m1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: nrxn1a
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001080021.2
NP_001073490.1
13 - 14
3449
87
XM_005156209.4
XP_005156266.1
13 - 14
3431
87
XM_009306641.3
XP_009304916.1
13 - 14
3431
87
XM_009306642.3
XP_009304917.1
13 - 14
3428
87
XM_009306643.3
XP_009304918.1
13 - 14
3430
87
XM_009306644.3
XP_009304919.1
13 - 14
3431
87
XM_009306645.3
XP_009304920.1
12 - 13
3419
87
XM_009306646.3
XP_009304921.1
12 - 13
3416
87
XM_009306648.3
XP_009304923.1
12 - 13
3413
87
XM_009306649.3
XP_009304924.1
12 - 13
3407
87
XM_009306650.3
XP_009304925.1
12 - 13
3404
87
XM_009306651.3
XP_009304926.1
13 - 14
3430
87
XM_009306652.3
XP_009304927.1
13 - 14
3431
87
XM_009306653.3
XP_009304928.1
13 - 14
3431
87
XM_009306654.3
XP_009304929.1
13 - 14
3431
87
XM_009306656.3
XP_009304931.1
13 - 14
3431
87
XM_021480136.1
XP_021335811.1
11 - 12
3401
87
XM_021480137.1
XP_021335812.1
13 - 14
3430
87
XM_021480138.1
XP_021335813.1
10 - 11
3371
87
XM_021480139.1
XP_021335814.1
12 - 13
3404
87
XM_021480140.1
XP_021335815.1
11 - 12
3394
87
XM_021480141.1
XP_021335816.1
12 - 13
3404
87
XM_021480142.1
XP_021335817.1
8 - 9
3320
87
XM_021480143.1
XP_021335818.1
11 - 12
3400
87
XM_021480144.1
XP_021335819.1
9 - 10
3343
87
XM_021480145.1
XP_021335820.1
8 - 9
3320
87
XM_021480146.1
XP_021335821.1
9 - 10
3344
87
GenBank mRNA
DQ641424.1
-
12 - 13
3398
87
Back To Top
More Information
Set Membership:
Amplicon length between 86 and 100
Danio rerio
Probe spans exons
Gene Ontology Categories:
Function(s)
Process(es)
sprouting angiogenesis
cell adhesion
metal ion binding
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-r642z:80/10.239.167.51:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0