Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sept8b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083566.1 | NP_001077035.1 | 9 - 10 | 1271 | 75 | |
XM_005169631.3 | XP_005169688.1 | 10 - 11 | 1397 | 75 | ||
XM_021481055.1 | XP_021336730.1 | 9 - 10 | 1254 | 75 | ||
GenBank mRNA | BC115182.1 | - | 9 - 10 | 1271 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |