Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: aldoab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213215.1 | NP_998380.1 | 6 - 7 | 1020 | 85 | |
GenBank mRNA | BC065847.1 | - | 6 - 7 | 1020 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |