Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cnot8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213479.1 | NP_998644.1 | 4 - 5 | 587 | 64 | |
XM_005173877.3 | XP_005173934.1 | 4 - 5 | 705 | 64 | ||
GenBank mRNA | BC055263.1 | - | 4 - 5 | 587 | 64 | |
BC153627.1 | - | 4 - 5 | 599 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |