Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fgfrl1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200376.1 | NP_956670.1 | 5 - 6 | 796 | 69 | |
GenBank mRNA | AJ574916.1 | - | 5 - 6 | 797 | 69 | |
BC053245.1 | - | 5 - 6 | 796 | 69 | ||
CU458922.1 | - | 5 - 6 | 797 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |